missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Magnesium hydroxide, 95%, pure, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 315720025

Catalog No. AC315720025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Magnesium hydroxide
1309-42-8
94% min. (Complexometry)
Mg(OH)2
15, 5734
Solubility in water: insoluble
[OH-].[OH-].[Mg+2]
58.33
CHEBI:6637
95%
Plastic bottle
95%
2.5kg
H2O2Mg
MFCD00011104
milk of magnesia, magnesium dihydroxide, brucite, magnesium hydroxide suspension, magnesium hydroxide gilumag, magnesiumhydroxid, magnesiumhydroxide, magnesia,, acmc-1bqga, magnesium 2+ hydroxide
VTHJTEIRLNZDEV-UHFFFAOYSA-L
magnesium;dihydroxide
73981
58.33
Pure
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

AIR SENSITIVE

EINECSNumber : 215-170-3

RTECSNumber : OM3570000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only