missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Magnesium pyrophosphate, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 03931730

 View more versions of this product

Catalog No. AA3931730


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Magnesium pyrophosphate
13446-24-7
250 g
MFCD00049482
magnesium pyrophosphate, unii-al34y660jv, diphosphoric acid, magnesium salt 1:2, dimagnesium diphosphate, acmc-1but0, 2mg.p2o7, magnesium pyrophosphate 50g, dimagnesium phosphonato phosphate, dimagnesium 2+ diphosphate, dimagnesium 2+ ion diphosphate
XZTWHWHGBBCSMX-UHFFFAOYSA-J
dimagnesium;phosphonato phosphate
122213
White
Powder
Mg2O7P2
14,5684
Soluble in dilute mineral acids. Insoluble in water.
[O-]P(=O)([O-])OP(=O)([O-])[O-].[Mg+2].[Mg+2]
222.551
222.57

Chemical Identifiers

13446-24-7
222.551
XZTWHWHGBBCSMX-UHFFFAOYSA-J
122213
[O-]P(=O)([O-])OP(=O)([O-])[O-].[Mg+2].[Mg+2]
Mg2O7P2
MFCD00049482
magnesium pyrophosphate, unii-al34y660jv, diphosphoric acid, magnesium salt 1:2, dimagnesium diphosphate, acmc-1but0, 2mg.p2o7, magnesium pyrophosphate 50g, dimagnesium phosphonato phosphate, dimagnesium 2+ diphosphate, dimagnesium 2+ ion diphosphate
dimagnesium;phosphonato phosphate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 236-595-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only