Learn More
Magnesium, Reagent, Ribbon, +99%, Thermo Scientific Chemicals
$167.38 - $167.38
Chemical Identifiers
CAS | 7439-95-4 |
---|---|
Molecular Formula | Mg |
Molecular Weight (g/mol) | 24.30 |
MDL Number | MFCD00085308 |
InChI Key | JLVVSXFLKOJNIY-UHFFFAOYSA-N |
Synonym | magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs |
PubChem CID | 5462224 |
ChEBI | CHEBI:25107 |
IUPAC Name | magnesium |
SMILES | [Mg] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC413380250
|
Thermo Scientific Chemicals
413380250 |
25 g |
Each for $167.38
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7439-95-4 | |
24.30 | |
JLVVSXFLKOJNIY-UHFFFAOYSA-N | |
5462224 | |
magnesium |
Mg | |
MFCD00085308 | |
magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs | |
CHEBI:25107 | |
[Mg] |
Specifications
White | |
7439-95-4 | |
1.74 | |
Mg | |
MFCD00085308 | |
25 g | |
500°C | |
magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs | |
[Mg] | |
24.30 | |
CHEBI:25107 | |
≥99% | |
Magnesium, 99+% |
651.0°C | |
1107.0°C | |
99+% | |
Mg | |
1869 | |
01,627; 02,254; 03,189; 04,315; 05,419; 06,351; 07,218; 10,251; 12,290; 13,170; 15,120; 16,198 | |
15, 5717 | |
JLVVSXFLKOJNIY-UHFFFAOYSA-N | |
magnesium | |
5462224 | |
24.3 | |
Ribbon |
Safety and Handling
GHS H Statement:
In contact with water releases flammable gases.
Self-heating in large quantities; may catch fire.
Flammable solid.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep away from any possible contact with water, because of violent reaction and possible flash fire.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
EINECSNumber : 231-104-6
TSCA : TSCA
RUO – Research Use Only