missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Magnesium ribbon, approximate dimensions, 3.0mm (0.12in) wide, 0.15mm (0.0059in) thick, Thermo Scientific Chemicals

Form: approx. 0.78g/m; Highly flammable

$218.58 - $924.29

Chemical Identifiers

CAS 7439-95-4
Molecular Formula Mg
Molecular Weight (g/mol) 24.30
MDL Number MFCD00085308
InChI Key JLVVSXFLKOJNIY-UHFFFAOYSA-N
Synonym magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs
PubChem CID 5462224
ChEBI CHEBI:25107
IUPAC Name magnesium
SMILES [Mg]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1281114
View Documents
Thermo Scientific Chemicals
01281114
25 g
Each for $218.58
 
AA1281151
View Documents
Thermo Scientific Chemicals
01281151
5 x 25 g
Each for $924.29
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7439-95-4
24.30
JLVVSXFLKOJNIY-UHFFFAOYSA-N
5462224
magnesium
Mg
MFCD00085308
magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs
CHEBI:25107
[Mg]

Specifications

7439-95-4
MFCD00085308
25 g
JLVVSXFLKOJNIY-UHFFFAOYSA-N
magnesium
5462224
Odorless
Magnesium ribbon, Approximate dimensions: 3.0mm (0.12 in.) wide, 0.15mm (0.0059 in.) thick
Mg
UN1869
magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs
[Mg]
24.30
CHEBI:25107
≈0.78g/m
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228-H251-H261
Flammable solid.
Self-heating: may catch fire.
In contact with water releases flammable gas.

P210-P223-P231+P232-P235+P410-P240-P241-P280-P335+P334-P370+P378s-P501c

H228-H252-H261

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: MAGNESIUM

EINECSNumber : 231-104-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only