Learn More
Magnesium silicate monohydrate (Talc), Thermo Scientific Chemicals
Lubricant, Filler, Electric and Heat Insulator
$46.99 - $272.71
Chemical Identifiers
CAS | 14807-96-6 |
---|---|
Molecular Formula | H2Mg3O12Si4 |
Molecular Weight (g/mol) | 379.26 |
MDL Number | MFCD00792903 |
InChI Key | KNXVOGGZOFOROK-UHFFFAOYSA-N |
Synonym | magnesium silicate, hydrous, talc powder, talc, cp, talc,-350 mesh, talc, powder, 10 mum, 3mgo.4o2si.h2o, talc, vetec tm reagent grade, talc, tested according to ph.eur., magnesium silicate monohydrate talc, tris @magnesium oxide tetrakis silica hydrate |
PubChem CID | 16211421 |
IUPAC Name | dioxosilane;oxomagnesium;hydrate |
SMILES | [Mg++].[Mg++].[Mg++].O[Si]([O-])=O.O[Si]([O-])=O.[O-][Si]([O-])=O.[O-][Si]([O-])=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA4031822
|
Thermo Scientific Chemicals
04031822 |
100 g |
Each for $46.99
|
|
|||||
AA4031836
|
Thermo Scientific Chemicals
04031836 |
500 g |
Each for $93.51
|
|
|||||
AA40318A4
|
Thermo Scientific Chemicals
040318A4 |
2.5 kg |
Each for $272.71
|
|
|||||
Description
Magnesium silicate is used as a lubricant, filler, electric and heat insulator and in clarifying liquids by filtration. Talc is used in many industries such as paper making, plastic, paints and coatings, rubber, food, electric cable, pharmaceuticals, cosmetics and ceramics. It is a widely used as baby powder (aka talcum). It is also useful for the treatment of complicated spontaneous pneumothorax.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14807-96-6 | |
379.26 | |
KNXVOGGZOFOROK-UHFFFAOYSA-N | |
16211421 | |
[Mg++].[Mg++].[Mg++].O[Si]([O-])=O.O[Si]([O-])=O.[O-][Si]([O-])=O.[O-][Si]([O-])=O |
H2Mg3O12Si4 | |
MFCD00792903 | |
magnesium silicate, hydrous, talc powder, talc, cp, talc,-350 mesh, talc, powder, 10 mum, 3mgo.4o2si.h2o, talc, vetec tm reagent grade, talc, tested according to ph.eur., magnesium silicate monohydrate talc, tris @magnesium oxide tetrakis silica hydrate | |
dioxosilane;oxomagnesium;hydrate |
Specifications
Gray to White | |
Powder | |
H2Mg3O12Si4 | |
magnesium silicate, hydrous, talc powder, talc, cp, talc,-350 mesh, talc, powder, 10 mum, 3mgo.4o2si.h2o, talc, vetec tm reagent grade, talc, tested according to ph.eur., magnesium silicate monohydrate talc, tris @magnesium oxide tetrakis silica hydrate | |
KNXVOGGZOFOROK-UHFFFAOYSA-N | |
dioxosilane;oxomagnesium;hydrate | |
16211421 | |
Odorless | |
2.7 g/cm3 at 20°C |
14807-96-6 | |
100 g | |
MFCD00792903 | |
Insoluble in water,cold acids,alkalies. | |
[Mg++].[Mg++].[Mg++].O[Si]([O-])=O.O[Si]([O-])=O.[O-][Si]([O-])=O.[O-][Si]([O-])=O | |
379.26 | |
379.28 (361.27 Anhydrous) | |
White to pale gray powder | |
Magnesium silicate monohydrate (Talc) |
Safety and Handling
P201-P202-P281-P308+P313-P501c
H350
EINECSNumber : 238-877-9
RTECSNumber : WW2710000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only