missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Magnesium sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.95% (metals basis), Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 40861KS

Catalog No. AA40861KS


Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Magnesium sputtering target
7439-95-4
Mg
1 Ea.
JLVVSXFLKOJNIY-UHFFFAOYSA-N
magnesium
5462224
99.95%
76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick
(metals basis)
MFCD00085308
magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs
[Mg]
24.30
CHEBI:25107
Odorless

Chemical Identifiers

7439-95-4
24.30
JLVVSXFLKOJNIY-UHFFFAOYSA-N
5462224
magnesium
Mg
MFCD00085308
magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs
CHEBI:25107
[Mg]
Videos
Safety and Handling

Safety and Handling

P210-P223-P231+P232-P235+P410-P240-P241-P280-P335+P334-P370+P378s-P501c

H228-H252-H261

DOTInformation : Hazard Class: 4.1; Packaging Group: III

EINECSNumber : 231-104-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions