missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Magnesium Standard, 1000mg/L in 2% HNO3 (Assurance Grade), SPEX CertiPrep

Supplier:  SPEX CertiPrep Group LLC PLMG22T

 View more versions of this product

Catalog No. PLMG22T


Explore available promotions

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

  • Magnesium Standard
  • Made with ASTM Type I Water
  • 1,000 μg/mL concentration
  • 250 mL volume

Certifications

ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015

Specifications

Specifications

Magnesium Standard
Poly Bottle
Assurance Certificate of Analysis reports up to 30 elements at ppb levels.
MFCD00085308
[Mg]
24.30
CHEBI:25107
7439-95-4,7697-37-2,7732-18-5
250 mL
Mg
JLVVSXFLKOJNIY-UHFFFAOYSA-N
magnesium
5462224
Assurance

Chemical Identifiers

7439-95-4
24.30
JLVVSXFLKOJNIY-UHFFFAOYSA-N
CHEBI:25107
[Mg]
Mg
MFCD00085308
5462224
magnesium
Safety and Handling

Safety and Handling

DOTInformation : Nonhazardous

Recommended Storage : Ambient

SDS
Documents

Documents

Promotions

Promotions