missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Magnesium stearate, 3.8-5.0% Mg, Thermo Scientific Chemicals
$68.09 - $352.58
Chemical Identifiers
CAS | 557-04-0 |
---|---|
Molecular Formula | C36H70MgO4 |
Molecular Weight (g/mol) | 591.27 |
InChI Key | HQKMJHAJHXVSDF-UHFFFAOYSA-L |
Synonym | magnesium stearate, magnesium octadecanoate, magnesium distearate, synpro 90, octadecanoic acid, magnesium salt, dibasic magnesium stearate, petrac mg 20nf, ns-m salt, sm-p, stearic acid, magnesium salt |
PubChem CID | 11177 |
ChEBI | CHEBI:9254 |
IUPAC Name | magnesium;octadecanoate |
SMILES | CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-].[Mg+2] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC240760250
|
Thermo Scientific Chemicals
240760250 |
25 g | Plastic bottle |
Each for $68.09
|
|
|||||
AC240760010
|
Thermo Scientific Chemicals
240760010 |
1 kg | Plastic bottle |
Each for $89.96
|
|
|||||
AC240760050
|
Thermo Scientific Chemicals
240760050 |
5 kg | Plastic drum |
Each for $352.58
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
557-04-0 | |
591.27 | |
magnesium stearate, magnesium octadecanoate, magnesium distearate, synpro 90, octadecanoic acid, magnesium salt, dibasic magnesium stearate, petrac mg 20nf, ns-m salt, sm-p, stearic acid, magnesium salt | |
CHEBI:9254 | |
CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-].[Mg+2] |
C36H70MgO4 | |
HQKMJHAJHXVSDF-UHFFFAOYSA-L | |
11177 | |
magnesium;octadecanoate |
Specifications
557-04-0 | |
White | |
Authentic | |
C36H70MgO4 | |
25 g | |
magnesium stearate, magnesium octadecanoate, magnesium distearate, synpro 90, octadecanoic acid, magnesium salt, dibasic magnesium stearate, petrac mg 20nf, ns-m salt, sm-p, stearic acid, magnesium salt | |
HQKMJHAJHXVSDF-UHFFFAOYSA-L | |
magnesium;octadecanoate | |
11177 | |
591.27 | |
Fine Powder |
130.0°C to 140.0°C | |
6% (1g, 105°C) max. | |
Plastic bottle | |
Mg(C18H35O2)2 | |
15,5754 | |
Solubility in water: insoluble. Other solubilities: decomposes in diluted acids,practically insoluble in alcohols and ether | |
CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-].[Mg+2] | |
591.27 | |
CHEBI:9254 | |
3.8-5% Mg | |
Magnesium stearate |
Safety and Handling
EINECSNumber : 209-150-3
RUO – Research Use Only