missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Magnesium sulfate, 97%, pure, anhydrous, Thermo Scientific Chemicals
Magnesium sulfate, 97%, MgO4S, CAS Number-7487-88-9, sulfuric acid magnesium salt 1:1, sal de sedlitz, magnesium sulfate, tomix ot, bitter salt, magnesium sulfate 1:1, sal angalis, magnesium sulphate, mgso4, magnesium sulfate anhydrous, 500g, CHEBI:32599, Undesignated, >700 deg.C
Supplier: Thermo Scientific Chemicals 413485000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
AdsorbentSpecifications
Magnesium sulfate | |
Pure | |
Granular Powder and/or Specks | |
97% | |
MgSO4 | |
01,634; 04,425; 05,421 | |
magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot | |
CSNNHWWHGAXBCP-UHFFFAOYSA-L | |
magnesium;sulfate | |
24083 | |
120.37 | |
Pure | |
>700°C |
1124.0°C | |
7487-88-9 | |
500 g | |
MgO4S | |
MFCD00011110 | |
15, 5755 | |
Solubility in water: 26g/100mL (0°C). | |
[Mg++].[O-]S([O-])(=O)=O | |
120.36 | |
CHEBI:32599 | |
97% | |
Plastic bottle |
Chemical Identifiers
7487-88-9 | |
120.36 | |
CSNNHWWHGAXBCP-UHFFFAOYSA-L | |
24083 | |
magnesium;sulfate |
MgO4S | |
MFCD00011110 | |
magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot | |
CHEBI:32599 | |
[Mg++].[O-]S([O-])(=O)=O |
RUO – Research Use Only