missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing

Magnesium Sulfate Anhydrous (Powder/Certified), Fisher Chemical™
$333.75 - $12020.62
Chemical Identifiers
CAS | 7487-88-9 |
---|---|
Molecular Formula | MgO4S |
Molecular Weight (g/mol) | 120.36 |
MDL Number | MFCD00011110 |
InChI Key | CSNNHWWHGAXBCP-UHFFFAOYSA-L |
Synonym | magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot |
PubChem CID | 24083 |
ChEBI | CHEBI:32599 |
IUPAC Name | magnesium(2+) sulfate |
SMILES | [Mg++].[O-]S([O-])(=O)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
M65-500
![]() |
Fisher Chemical
M65500 |
500 g | Poly Bottle |
Each for $333.75
Case of 6 Each for $2,002.50
|
![]() |
|||||
M65-100LB
|
Fisher Chemical
FLM65100LB |
100 lb. | Fiber Drum |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | ||||||||||
M65-3
|
Fisher Chemical
M653 |
3 kg | Poly Bottle |
Each for $1,057.58
Case of 4 Each for $4,230.32
|
|
|||||
Chemical Identifiers
7487-88-9 | |
120.36 | |
CSNNHWWHGAXBCP-UHFFFAOYSA-L | |
24083 | |
magnesium(2+) sulfate |
MgO4S | |
MFCD00011110 | |
magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot | |
CHEBI:32599 | |
[Mg++].[O-]S([O-])(=O)=O |
Specifications
1124°C | |
White | |
Solid | |
MgO4S | |
magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot | |
[Mg++].[O-]S([O-])(=O)=O | |
120.36 | |
CHEBI:32599 | |
Certified | |
Poly Bottle | |
2.65g/cm³ |
7487-88-9 | |
7.9 | |
500 g | |
MFCD00011110 | |
CSNNHWWHGAXBCP-UHFFFAOYSA-L | |
magnesium(2+) sulfate | |
24083 | |
120.37 | |
Pass Test | |
>700°C | |
Magnesium Sulfate Anhydrous |
Safety and Handling

CAUTION!
Emergency Overview
May cause eye, skin, and respiratory tract irritation. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.
NFPA
Health:1
Flammability:0
Instability:1