missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Magnesium sulfate, dried, contains ca 1-2 mol water of hydration, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L137390E
Description
Suitable for the preliminary drying of organic liquids, for example solvent extracts from reaction workups.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Magnesium sulfate | |
22189-08-8 | |
MgO4S | |
14,5691 | |
CSNNHWWHGAXBCP-UHFFFAOYSA-L | |
magnesium;sulfate | |
24083 | |
120.37 (Anhydrous) |
contains ≈1 to 2 mol water of hydration | |
2500 g | |
MFCD00149787 | |
magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot | |
[Mg++].[O-]S([O-])(=O)=O | |
120.36 | |
CHEBI:32599 | |
Hygroscopic |
Chemical Identifiers
22189-08-8 | |
120.36 | |
CSNNHWWHGAXBCP-UHFFFAOYSA-L | |
24083 | |
magnesium;sulfate |
MgO4S | |
MFCD00149787 | |
magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot | |
CHEBI:32599 | |
[Mg++].[O-]S([O-])(=O)=O |
Safety and Handling
EINECSNumber : 231-298-2
RTECSNumber : OM4500000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only