missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Magnesium Sulfate Heptahydrate, ≥99%, MP Biomedicals

Supplier:  MP Biomedicals Inc 0219483391

 View more versions of this product

Catalog No. ICN-19483391

Explore available promotions

Description

Description

Specifications

Specifications

Magnesium Sulfate Heptahydrate
1 kg
H14MgO11S
magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate
O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O
246.47
CHEBI:31795
10034-99-8
≥99%
MFCD00149785
WRUGWIBCXHJTDG-UHFFFAOYSA-L
magnesium(2+) heptahydrate sulfate
24843
246.5

Chemical Identifiers

10034-99-8
246.47
WRUGWIBCXHJTDG-UHFFFAOYSA-L
24843
magnesium(2+) heptahydrate sulfate
H14MgO11S
MFCD00149785
magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate
CHEBI:31795
O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions