missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Magnesium Sulfate Solution (2.25%/APHA), Fisher Chemical
Supplier: Thermo Fisher Scientific SM1091
Description
EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.
Specifications
Clear | |
Glass Bottle | |
Magnesium Sulfate Heptahydrate Solution, 2.25% |
Liquid | |
1 L |