missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Magnesium trisilicate, Thermo Scientific Chemicals
$95.19 - $144.64
Chemical Identifiers
CAS | 14987-04-3 |
---|---|
Molecular Formula | Mg2O8Si3 |
MDL Number | MFCD00016203 |
Synonym | Magnesium silicate |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14987-04-3 | |
Mg2O8Si3 | |
MFCD00016203 | |
Magnesium silicate |
Specifications
White | |
Powder | |
Mg2O8Si3 | |
Magnesium silicate | |
Magnesium trisilicate |
14987-04-3 | |
250 g | |
MFCD00016203 | |
260.86 |
Safety and Handling
EINECSNumber : 239-076-7
RTECSNumber : IN6620000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only