missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Magnesium, turnings, 99+%, Thermo Scientific Chemicals

$76.52 - $608.12

Chemical Identifiers

CAS 7439-95-4
Molecular Formula Mg
Molecular Weight (g/mol) 24.30
MDL Number MFCD00085308
InChI Key JLVVSXFLKOJNIY-UHFFFAOYSA-N
Synonym magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs
PubChem CID 5462224
ChEBI CHEBI:25107
IUPAC Name magnesium
SMILES [Mg]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0812022
View Documents
Thermo Scientific Chemicals
L0812022
100 g
Each for $76.52
 
AAL0812036
View Documents
Thermo Scientific Chemicals
L0812036
500 g
Each for $165.44
 
AAL081200E
View Documents
Thermo Scientific Chemicals
L081200E
2500 g
Each for $608.12
 
Description

Description

Magnesium, turnings is used in solid state synthesis with Ca and Sn resulted in a new phase, Ca6.2Mg3.8Sn7, which has an unprecedented type of tin chain composed of square-planar tin units. It is a component in die-casting, to remove sulfur in the production of iron and steel, the production of titanium in the Kroll process. It is widely used for manufacturing of mobile phones, laptop and tablet computers, cameras, and other electronic components.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Magnesium, turnings is used in solid state synthesis with Ca and Sn resulted in a new phase, Ca6.2Mg3.8Sn7, which has an unprecedented type of tin chain composed of square-planar tin units. It is a component in die-casting, to remove sulfur in the production of iron and steel, the production of titanium in the Kroll process. It is widely used for manufacturing of mobile phones, laptop and tablet computers, cameras, and other electronic components.

Solubility
Reacts with water.

Notes
Store away from air, moisture and oxidizing agents. Incompatible with acids, air, nitrogen and moisture.
Specifications

Chemical Identifiers

7439-95-4
24.30
JLVVSXFLKOJNIY-UHFFFAOYSA-N
5462224
magnesium
Mg
MFCD00085308
magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs
CHEBI:25107
[Mg]

Specifications

650°C
1091°CC
Mg
UN1869
4948473
Reacts with water.
[Mg]
24.30
CHEBI:25107
≥99%
Moisture sensitive
7439-95-4
1.738
MFCD00085308
100 g
magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs
JLVVSXFLKOJNIY-UHFFFAOYSA-N
magnesium
5462224
24.305
Odorless
Magnesium, turnings
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228-H251-H261
Flammable solid.
Self-heating: may catch fire.
In contact with water releases flammable gas.

P210-P223-P231+P232-P235+P410-P240-P241-P280-P335+P334-P370+P378s-P501c

H228-H252-H261

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: MAGNESIUM

EINECSNumber : 231-104-6

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only