Learn More
Eisco™ Magnetic Field Demonstrator
With two transparent plates to demonstrate magnet fields.
Supplier: Eisco™ PH0802MF2
Description
- Set of two transparent plastic plates
- Each plate has matrix of 98 (14 x 7) small circular chambers
- Each chamber contains iron rod functioning as plotting compass
- Plates may be arranged as a single large two-dimensional surface, as faces of cube, etc.
- Transparent nature of plates enables for use on overhead projector
Specifications
| Plate with matrix of 98 (14 x 7) small circular chambers | |
| 9 to 12 | |
| 1.2 in. | |
| 3 cm | |
| 16 cm | |
| 3.1 in. | |
| 0.3 lb. | |
| 15.3 x 7.6 x 0.6 cm |
| Demonstrate magnet fields | |
| Magnetic Field Demonstrator | |
| Two transparent plates | |
| 6.3 in. | |
| 136 g | |
| 15.3 x 7.6 x 0.6cm | |
| 7.87 cm |