Supertek™ Magnifier, Folding 10X DFS Item
Pocket type magnifier mounted in single body with a metal cover which doubles as a handle fitted with a single lens of diameter 18 mm with a magnification of x 10.
Supplier:  Supertek™ PH30186
Catalog No. S012325
Explore available promotions
Description
Description
  • Pocket type magnifier.
  • Mounted in single body with a metal cover which doubles as a handle.
  • Single 18 mm diamer lens.
  • 10X magnification.
Specifications
Specifications
18 mm
Glass, Metal
10x
Magnifier
5/8 in.