Carson™ MagniLoupe Eye Loupe Magnifier Set with Smartphone Digiscoping Clip DFS Item
Carson™ MagniLoupe Eye Loupe Magnifier Set includes 4 Magnification Options: 4.5X, 6.5X, 8X and 13X.
Supplier:  Carson™ ML20
Catalog No. S81149
Explore available promotions
Description
Description
  • Contains 4 ergonomically designed eye loupe magnifiers that are engineered to rest comfortably in the eye socket
  • 4 different magnifications: 4.5X, 6.5X, 8X and 13X powers.
  • Distortion-free acrylic lens design is extremely compact and lightweight
  • Smartphone digiscoping clip that attaches to each of the loupes by removing the rubber eyecup and sliding the smartphone adapter clip onto the loupe until it clicks into place.
  • Smartphone digiscoping clip is compatible with iPhone, Samsung, LG, Huawei and Google Pixel phone models
  • Digiscoping Adapter is compatible with most cases, but it does not work on the largest Otter Box style cases
  • Excellent for a variety of hobbies, crafts and tasks or for jewelers, watchmakers, viewing printing pixels, rocks, stones, gems, stamps, currency, model building and other hobby, craft and inspection purposes
  • Limited Lifetime Warranty
Specifications
Specifications
4.5x/6.5x/8x/13x
Jewelers, Watchmakers, Currency, Inspection and More
Smartphone Digiscoping Clip