Invitrogen™ MagniSort™ Magnet
Designed to be used with 12 x 75mm round bottom polystyrene tubes (5mL), and can be used to magnetically separate up to 2 x 108 cells. For detailed instructions, follow the protocols provided with each separation kit.
Supplier: Invitrogen™ MAG490210
Alert:
This magnet generates a strong magnetic field. Keep away from pacemakers, credit cards, magnetic I.D. cards, watches, computer monitors and hard disks to prevent damage to these devices.
Description
This magnet is composed of strong rare earth magnets. Rare earth magnets are sensitive to heat above 175°F (80°C). Keep away from extreme heat source.Specifications
Magnet | |
5 mL | |
12 x 75 mm round bottom polystyrene tubes |
For Research Use Only. Not for use in diagnostic procedures.