Thermo Scientific™ Male Luer-LOK Priming Syringes
Thermo Scientific™ Male Luer-LOK Priming Syringes offer easy, accurate and reproducible manual injections.
$223.53 - $919.36
Products 2
Catalog Number Mfr. No. Volume (Metric) Product Type Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Product Type Price Quantity  
36-5KL-531
View Documents Promotion Details
Thermo Scientific™
365KL531
1 mL Gas Tight GC Autosampler Syringe, Luer-Lok
Each for $223.53
365N2721
View Documents Promotion Details
Thermo Scientific™
365N2721
10 mL GC Autosampler Syringe
Each for $919.36
Description
Description
Thermo Scientific™ Male Luer-LOK Priming Syringes offer easy, accurate and reproducible manual injections.
  • Square tip to prevent damage to the injector
  • Wide range of volumes
  • Precision made from borosilicate glass and stainless steel
  • Robust design and easy-to-read markings
SDS
missing translation for 'documents'
missing translation for 'documents'
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.