missing translation for 'onlineSavingsMsg'
Learn More

Maleic acid, 98+%

CAS: 110-16-7 | C4H4O4 | 116.07 g/mol

$55.60 - $488.99

Chemical Identifiers

CAS 110-16-7
Molecular Formula C4H4O4
Molecular Weight (g/mol) 116.07
MDL Number MFCD00063177
InChI Key VZCYOOQTPOCHFL-OWOJBTEDSA-N
Synonym maleic acid, cis-butenedioic acid, toxilic acid, maleinic acid, malenic acid, 2z-but-2-enedioic acid, 2-butenedioic acid z, maleate, cis-1,2-ethylenedicarboxylic acid, z-butenedioic acid
PubChem CID 444266
ChEBI CHEBI:18300
IUPAC Name (Z)-but-2-enedioic acid
SMILES OC(=O)\C=C\C(O)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1459630
View Documents
Thermo Scientific Chemicals
A1459630
250 g
Each for $55.60
Only null left
 
AAA145960B
View Documents
Thermo Scientific Chemicals
A145960B
1000 g
Each for $135.40
Only null left
 
AAA145960I
View Documents
Thermo Scientific Chemicals
A145960I
5000 g
Each for $488.99
Only null left
 
Description

Description

Maleic acid is used as a precursor to fumaric acid, dimethyl maleate and glyoxalic acid. It is an electrophile and acts as dienophine in Diels-Alder reactions. It reacts with drugs to form more stable addition salts like indacaterol maleate, carfenazine, chlorpheniramine, pyrilamine, methylergonovine and thiethylperazine. Its maleate ion is useful in biochemistry as an inhibitor of transaminase reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Maleic acid is used as a precursor to fumaric acid, dimethyl maleate and glyoxalic acid. It is an electrophile and acts as dienophine in Diels-Alder reactions. It reacts with drugs to form more stable addition salts like indacaterol maleate, carfenazine, chlorpheniramine, pyrilamine, methylergonovine and thiethylperazine. Its maleate ion is useful in biochemistry as an inhibitor of transaminase reactions.

Solubility
Soluble in water, acetone, glacial acetic acid, alcohol and concentrated sulfuric acid. Slightly soluble in diethyl ether. Insoluble in benzene and chloroform.

Notes
Incompatible with oxidizing agents, bases, reducing agents, metals and alkalis.
Specifications

Chemical Identifiers

110-16-7
116.07
VZCYOOQTPOCHFL-OWOJBTEDSA-N
444266
(Z)-but-2-enedioic acid
C4H4O4
MFCD00063177
maleic acid, cis-butenedioic acid, toxilic acid, maleinic acid, malenic acid, 2z-but-2-enedioic acid, 2-butenedioic acid z, maleate, cis-1,2-ethylenedicarboxylic acid, z-butenedioic acid
CHEBI:18300
OC(=O)\C=C\C(O)=O

Specifications

132°C to 136°C
C4H4O4
605762
maleic acid, cis-butenedioic acid, toxilic acid, maleinic acid, malenic acid, 2z-but-2-enedioic acid, 2-butenedioic acid z, maleate, cis-1,2-ethylenedicarboxylic acid, z-butenedioic acid
VZCYOOQTPOCHFL-OWOJBTEDSA-N
(Z)-but-2-enedioic acid
444266
116.07
1.59
127°C (260°F)
110-16-7
MFCD00063177
14,5703
Soluble in water,acetone,glacial acetic acid,alcohol and concentrated sulfuric acid. Slightly soluble in diethyl ether. Insoluble in benzene and chloroform.
OC(=O)\C=C\C(O)=O
116.07
CHEBI:18300
≥98%
250 g
Maleic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H317-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c

H302+H312-H314-H317-H335

DOTInformation : Hazard Class: 8; Packaging Group: II

EINECSNumber : 203-742-5

RTECSNumber : OM9625000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only