Learn More
Maleic anhydride, 99%, pastilles
CAS: 108-31-6 | C4H2O3 | 98.06 g/mol
$52.67 - $866.34
Chemical Identifiers
| CAS | 108-31-6 |
|---|---|
| Molecular Formula | C4H2O3 |
| Molecular Weight (g/mol) | 98.06 |
| MDL Number | MFCD00005518 |
| InChI Key | FPYJFEHAWHCUMM-UHFFFAOYSA-N |
| Synonym | maleic anhydride, 2,5-furandione, maleic acid anhydride, toxilic anhydride, dihydro-2,5-dioxofuran, cis-butenedioic anhydride, 2,5-dihydrofuran-2,5-dione, polymaleic anhydride, maleinanhydrid, rcra waste number u147 |
| PubChem CID | 7923 |
| ChEBI | CHEBI:474859 |
| SMILES | O=C1OC(=O)C=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC125240250
|
Thermo Scientific Chemicals
125240250 |
25 g | Plastic bottle |
Each for $60.38
|
|
||||
|
AC125240010
|
Thermo Scientific Chemicals
125240010 |
1 kg | Plastic bottle |
Each for $52.67
|
|
||||
|
AC125240050
|
Thermo Scientific Chemicals
125240050 |
5 kg | Plastic drum |
Each for $183.49
|
|
||||
|
AC125240251
|
Thermo Scientific Chemicals
125240251 |
25 kg | Fibre drum |
Each for $866.34
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 108-31-6 | |
| 98.06 | |
| FPYJFEHAWHCUMM-UHFFFAOYSA-N | |
| 7923 | |
| O=C1OC(=O)C=C1 |
| C4H2O3 | |
| MFCD00005518 | |
| maleic anhydride, 2,5-furandione, maleic acid anhydride, toxilic anhydride, dihydro-2,5-dioxofuran, cis-butenedioic anhydride, 2,5-dihydrofuran-2,5-dione, polymaleic anhydride, maleinanhydrid, rcra waste number u147 | |
| CHEBI:474859 |
Specifications
| 108-31-6 | |
| White | |
| 200.0°C | |
| Authentic | |
| Plastic bottle | |
| MFCD00005518 | |
| 17, V,11, 55 | |
| 1.48 | |
| maleic anhydride, 2,5-furandione, maleic acid anhydride, toxilic anhydride, dihydro-2,5-dioxofuran, cis-butenedioic anhydride, 2,5-dihydrofuran-2,5-dione, polymaleic anhydride, maleinanhydrid, rcra waste number u147 | |
| FPYJFEHAWHCUMM-UHFFFAOYSA-N | |
| furan-2,5-dione | |
| 7923 | |
| CHEBI:474859 | |
| 99% | |
| Pellets |
| 52.0°C to 55.0°C | |
| 1.4800g/mL | |
| 102°C | |
| 99% | |
| C4H2O3 | |
| 25 g | |
| 04,316; 05,422 | |
| 15,5768 | |
| Solubility in water: 790g/L (25°C). Other solubilities: reacts with water forming maleic acid,227g/100g acetone- 50g/100g benzene (25°C),112g/100g ethylacetate (25°C)- 0,60g/100g ccl4,52.5g/100g chloroform (25°C)-23,4g/100g toluen | |
| O=C1OC(=O)C=C1 | |
| 98.06 | |
| 1.6 mPa.s (60°C) | |
| 98.06 | |
| Reagent | |
| Maleic anhydride |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes damage to organs through
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 203-571-6
RUO – Research Use Only