missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Malonic acid dihydrazide, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1227006

 View more versions of this product

Catalog No. AAL1227006

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Malonic acid dihydrazide
152°C to 156°C
MFCD00041268
1072337
PSIKPHJLTVSQFO-UHFFFAOYSA-N
propanedihydrazide
77433
99%
3815-86-9
C3H8N4O2
5 g
malonic acid dihydrazide, malonohydrazide, malonic dihydrazide, propanedioic acid, dihydrazide, malonhydrazide, malonyl dihydrazide, malonic acid, dihydrazide, malonyl hydrazide, malonic acid hydrazide, malonoyldihydrazine
NNC(=O)CC(=O)NN
132.12
132.12

Chemical Identifiers

3815-86-9
132.12
PSIKPHJLTVSQFO-UHFFFAOYSA-N
77433
NNC(=O)CC(=O)NN
C3H8N4O2
MFCD00041268
malonic acid dihydrazide, malonohydrazide, malonic dihydrazide, propanedioic acid, dihydrazide, malonhydrazide, malonyl dihydrazide, malonic acid, dihydrazide, malonyl hydrazide, malonic acid hydrazide, malonoyldihydrazine
propanedihydrazide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

RTECSNumber : OO8000000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only