missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Malonic acid disodium salt, 99%, Thermo Scientific Chemicals

$71.77 - $181.16

Chemical Identifiers

CAS 141-95-7
Molecular Formula C3H2Na2O4
Molecular Weight (g/mol) 148.025
MDL Number MFCD00002708
InChI Key PRWXGRGLHYDWPS-UHFFFAOYSA-L
Synonym sodium malonate, disodium malonate, propanedioic acid, disodium salt, malonic acid disodium salt, disodium propanedioate, malonic acid, sodium salt, unii-9qwe64m39h, propanedioic acid, sodium salt 1:2, malonic acid, disodium salt, dicarboxymethane disodium salt
PubChem CID 8865
ChEBI CHEBI:62983
IUPAC Name disodium;propanedioate
SMILES C(C(=O)[O-])C(=O)[O-].[Na+].[Na+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4484114
View Documents
Thermo Scientific Chemicals
04484114
25 g
Each for $71.77
 
AA4484122
View Documents
Thermo Scientific Chemicals
04484122
100 g
Each for $181.16
 
Description

Description

Malonic acid disodium salt is used in organic Synthesis, pharmaceuticals, agrochemicals and dye stuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

141-95-7
148.025
PRWXGRGLHYDWPS-UHFFFAOYSA-L
8865
disodium;propanedioate
C3H2Na2O4
MFCD00002708
sodium malonate, disodium malonate, propanedioic acid, disodium salt, malonic acid disodium salt, disodium propanedioate, malonic acid, sodium salt, unii-9qwe64m39h, propanedioic acid, sodium salt 1:2, malonic acid, disodium salt, dicarboxymethane disodium salt
CHEBI:62983
C(C(=O)[O-])C(=O)[O-].[Na+].[Na+]

Specifications

141-95-7
MFCD00002708
sodium malonate, disodium malonate, propanedioic acid, disodium salt, malonic acid disodium salt, disodium propanedioate, malonic acid, sodium salt, unii-9qwe64m39h, propanedioic acid, sodium salt 1:2, malonic acid, disodium salt, dicarboxymethane disodium salt
PRWXGRGLHYDWPS-UHFFFAOYSA-L
disodium;propanedioate
8865
148.03
Powder
C3H2Na2O4
25 g
Soluble in water (148g/L at 20°C).
C(C(=O)[O-])C(=O)[O-].[Na+].[Na+]
148.025
CHEBI:62983
99%
Malonic acid disodium salt
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 205-514-0

RTECSNumber : OO1750000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only