missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Malonic Acid, TCI America™

Supplier:  TCI America M002825G

 View more versions of this product

Catalog No. M002825G


Explore available promotions

Description

Description

Specifications

Specifications

Malonic Acid
134°C
C3H4O4
25 g
OFOBLEOULBTSOW-UHFFFAOYSA-N
propanedioic acid
867
104.06
Crystalline Powder
141-82-2
White
MFCD00002707
malonic acid, dicarboxymethane, carboxyacetic acid, methanedicarboxylic acid, kyselina malonova, usaf ek-695, dicarboxylate, dicarboxylic acid, kyselina malonova czech, propanediolic acid
C(C(=O)O)C(=O)O
104.061
CHEBI:30794
≥99.0% (T)

Chemical Identifiers

141-82-2
104.061
OFOBLEOULBTSOW-UHFFFAOYSA-N
867
propanedioic acid
C3H4O4
MFCD00002707
malonic acid, dicarboxymethane, carboxyacetic acid, methanedicarboxylic acid, kyselina malonova, usaf ek-695, dicarboxylate, dicarboxylic acid, kyselina malonova czech, propanediolic acid
CHEBI:30794
C(C(=O)O)C(=O)O
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0912

RTECSNumber : OO0175000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions