missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Maltotriose, 93%, Thermo Scientific Chemicals

$76.23 - $933.69

Chemical Identifiers

CAS 1109-28-0
MDL Number MFCD00006629
InChI Key FYGDTMLNYKFZSV-NBCPLHMPSA-N
Synonym maltotriose, d-maltotriose
PubChem CID 134129496
IUPAC Name (2R,3S,4S,5S,6R)-2-[(2R,3R,4R,5R,6R)-4,5-dihydroxy-2-(hydroxymethyl)-6-[(2R,3S,4S,5R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)OC3C(OC(C(C3O)O)O)CO)CO)O)O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC225945000
SDS View Documents
Thermo Scientific Chemicals
225945000
500 mg
Each for $76.23
 
AC225940250
SDS View Documents
Thermo Scientific Chemicals
225940250
25 g
Each for $933.69
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1109-28-0
FYGDTMLNYKFZSV-NBCPLHMPSA-N
134129496
C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)OC3C(OC(C(C3O)O)O)CO)CO)O)O)O)O
MFCD00006629
maltotriose, d-maltotriose
(2R,3S,4S,5S,6R)-2-[(2R,3R,4R,5R,6R)-4,5-dihydroxy-2-(hydroxymethyl)-6-[(2R,3S,4S,5R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol

Specifications

1109-28-0
White
91% min. (ELSD) (HPLC)
C18H32O16
500 mg
+ 162.50
Solubility in water: 50mg/mL,clear colorless solution
C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)OC3C(OC(C(C3O)O)O)CO)CO)O)O)O)O
504.44
504.44
Fine Crystalline Powder or Crystals
132.0°C to 135.0°C
Authentic
Glass bottle
MFCD00006629
+162.50° (25°C c=1,H2O)
maltotriose, d-maltotriose
FYGDTMLNYKFZSV-NBCPLHMPSA-N
(2R,3S,4S,5S,6R)-2-[(2R,3R,4R,5R,6R)-4,5-dihydroxy-2-(hydroxymethyl)-6-[(2R,3S,4S,5R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
134129496
93%
Maltotriose
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 214-174-2

SDS
Documents

Documents

RUO – Research Use Only