missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Mancozeb, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 468852500
Specifications
Mancozeb | |
Glass Bottle | |
Yellow to Gray | |
C8H12MnN4S8Zn | |
13-5738 | |
[Mn++].[Zn++].[S-]C(=S)NCCNC([S-])=S.[S-]C(=S)NCCNC([S-])=S | |
541.01 |
8018-01-7 | |
250 mg | |
Available | |
00078616 | |
CHNQZRKUZPNOOH-UHFFFAOYSA-J | |
manganese(2+) zinc(2+) bis(({2-[(sulfanidylmethanethioyl)amino]ethyl}carbamothioyl)sulfanide) | |
Powder |
Chemical Identifiers
8018-01-7 | |
541.01 | |
CHNQZRKUZPNOOH-UHFFFAOYSA-J | |
[Mn++].[Zn++].[S-]C(=S)NCCNC([S-])=S.[S-]C(=S)NCCNC([S-])=S |
C8H12MnN4S8Zn | |
00078616 | |
manganese(2+) zinc(2+) bis(({2-[(sulfanidylmethanethioyl)amino]ethyl}carbamothioyl)sulfanide) |
Safety and Handling
Health hazard
Exclamation mark
RTECSNumber : ZB3200000
Recommended Storage : Normal conditions