Learn More
Mandelic acid hydrazide, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1165306
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Mandelic acid hydrazide | |
128°C to 131°C | |
MFCD00038133 | |
2096197 | |
FWTGUGVETHVGTL-UHFFFAOYSA-N | |
2-hydroxy-2-phenylacetohydrazide | |
73126 | |
97% |
2443-66-5 | |
C8H10N2O2 | |
5 g | |
mandelic acid hydrazide, mandelhydrazide, mandelhydrazine, mandelic acid, hydrazide, mandelic hydrazide, hydroxy-phenyl-acetic acid hydrazide, 2-???-1-phenylethan-1-ol, mandelohydrazide, asischem d13400, akos bc-1667 | |
C1=CC=C(C=C1)C(C(=O)NN)O | |
166.18 | |
166.18 |
Chemical Identifiers
2443-66-5 | |
166.18 | |
FWTGUGVETHVGTL-UHFFFAOYSA-N | |
73126 | |
C1=CC=C(C=C1)C(C(=O)NN)O |
C8H10N2O2 | |
MFCD00038133 | |
mandelic acid hydrazide, mandelhydrazide, mandelhydrazine, mandelic acid, hydrazide, mandelic hydrazide, hydroxy-phenyl-acetic acid hydrazide, 2-???-1-phenylethan-1-ol, mandelohydrazide, asischem d13400, akos bc-1667 | |
2-hydroxy-2-phenylacetohydrazide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 000-000-0
RTECSNumber : OO7450000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only