missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Manganese, 99+%, powder, -40 mesh, Thermo Scientific Chemicals

$93.10 - $93.10

Chemical Identifiers

CAS 7439-96-5
Molecular Formula Mn
Molecular Weight (g/mol) 54.94
MDL Number MFCD00011111
InChI Key PWHULOQIROXLJO-UHFFFAOYSA-N
Synonym colloidal, mangan, manganese, elemental, cutaval, magnacat, metal alloy, tronamang, fume, mangan polish, compounds
PubChem CID 23930
ChEBI CHEBI:35154
IUPAC Name manganese
SMILES [Mn]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC317442500
SDS View Documents
Thermo Scientific Chemicals
317442500
250 g Glass bottle
Each for $93.10
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7439-96-5
54.94
PWHULOQIROXLJO-UHFFFAOYSA-N
23930
manganese
Mn
MFCD00011111
colloidal, mangan, manganese, elemental, cutaval, magnacat, metal alloy, tronamang, fume, mangan polish, compounds
CHEBI:35154
[Mn]

Specifications

Black-Brown to Brown-Gray
99+%
Mn
250 g
colloidal, mangan, manganese, elemental, cutaval, magnacat, metal alloy, tronamang, fume, mangan polish, compounds
[Mn]
54.94
CHEBI:35154
≥99.0% (Mn)
Powder
7439-96-5
Mn
MFCD00011111
15, 5788
PWHULOQIROXLJO-UHFFFAOYSA-N
manganese
23930
54.94
Glass bottle
Manganese, 99+%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Flammable solid.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 231-105-1

RTECSNumber : OO9275000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only