missing translation for 'onlineSavingsMsg'
Learn More

Manganese AA Standard, 1000 ppm Mn in 3% HCl, Ricca Chemical

$104.34 - $282.25

Chemical Identifiers

CAS 7439-96-5
Molecular Formula Mn
Molecular Weight (g/mol) 54.94
MDL Number MFCD00011111
InChI Key PWHULOQIROXLJO-UHFFFAOYSA-N
PubChem CID 23930
ChEBI CHEBI:35154
IUPAC Name manganese
SMILES [Mn]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AMN1KH-100
View Documents
Ricca Chemical Company
AMN1KH100
100 mL Natural LDPE Bottle
Each for $104.34
Only null left
 
AMN1KH-500
View Documents
Ricca Chemical Company
AMN1KH500
500 mL Natural Poly Bottle
Each for $282.25
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7439-96-5
54.94
PWHULOQIROXLJO-UHFFFAOYSA-N
CHEBI:35154
[Mn]
Mn
MFCD00011111
23930
manganese

Specifications

Natural LDPE Bottle
1,000ppm Mn
98.0,4.04,0.1
Liquid
MFCD00011111
100°C
[Mn]
54.94
23930
Manganese AA Standard
7439-96-5,7647-01-0,7732-18-5
94.0,3.88,0.09
1 mL = 1 mg Mn
Mn
Colorless
PWHULOQIROXLJO-UHFFFAOYSA-N
manganese
100 mL
CHEBI:35154
Videos
SDS
Documents

Documents