Learn More
Manganese(II) bromide, 99%, anhydrous
CAS: 13446-03-2 | Br2Mn | 214.76 g/mol
Supplier: Thermo Scientific Chemicals 207860250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Manganese(II) bromide | |
| Pink | |
| Powder | |
| 99% | |
| MnBr2 | |
| 15, 5790 | |
| 214.76 | |
| 99% | |
| 4.38 |
| Anhydrous | |
| 13446-03-2 | |
| 25 g | |
| Br2Mn | |
| MFCD00011113 | |
| Solubility in water: soluble | |
| 214.76 | |
| Glass bottle | |
| 4.3800g/mL |
Chemical Identifiers
| 13446-03-2 | |
| Br2Mn | |
| 214.76 | |
| MFCD00011113 |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS Signal Word: Warning
EINECSNumber : 236-591-9
RUO – Research Use Only