missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Manganese(II) chloride, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 271410025

 View more versions of this product

Catalog No. AC271410025


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1190.0°C
650.0°C
Pink
2.5 kg
Cl2Mn
MFCD00011114
15, 5793
Solubility in water: 723g/L (25°C). Other solubilities: soluble in alcohol,insoluble in ether
[Cl-].[Cl-].[Mn++]
125.84
CHEBI:63041
97%
Manganese(II) chloride
7773-01-5
Flakes
96% min. (Complexometry)
MnCl2
07,222
manganese chloride, manganese ii chloride, manganese 2+ ion dichloride, manganesechloride, acmc-20akkt, manganous chloride,anhydrous, ksc171o6h, manganous chloride, anhydr, manganese ii chloride anhydrous crystalline
GLFNIEUTAYBVOC-UHFFFAOYSA-L
manganese(2+) dichloride
10313134
125.84
Plastic Drum

Chemical Identifiers

7773-01-5
125.84
GLFNIEUTAYBVOC-UHFFFAOYSA-L
10313134
[Cl-].[Cl-].[Mn++]
Cl2Mn
MFCD00011114
manganese chloride, manganese ii chloride, manganese 2+ ion dichloride, manganesechloride, acmc-20akkt, manganous chloride,anhydrous, ksc171o6h, manganous chloride, anhydr, manganese ii chloride anhydrous crystalline
CHEBI:63041
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure if inhaled.
Causes serious eye damage.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Do not eat,drink or smoke when using th

GHS Signal Word: Danger

EINECSNumber : 231-869-6

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only