Learn More
Manganese(II) chloride, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 271412500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1190.0°C | |
650.0°C | |
Pink | |
250 g | |
Cl2Mn | |
MFCD00011114 | |
15, 5793 | |
Solubility in water: 723g/L (25°C). Other solubilities: soluble in alcohol,insoluble in ether | |
[Cl-].[Cl-].[Mn++] | |
125.84 | |
CHEBI:63041 | |
97% |
Manganese(II) chloride | |
7773-01-5 | |
Flakes | |
96% min. (Complexometry) | |
MnCl2 | |
07,222 | |
manganese chloride, manganese ii chloride, manganese 2+ ion dichloride, manganesechloride, acmc-20akkt, manganous chloride,anhydrous, ksc171o6h, manganous chloride, anhydr, manganese ii chloride anhydrous crystalline | |
GLFNIEUTAYBVOC-UHFFFAOYSA-L | |
manganese(2+);dichloride | |
10313134 | |
125.84 | |
Glass bottle |
Chemical Identifiers
7773-01-5 | |
125.84 | |
GLFNIEUTAYBVOC-UHFFFAOYSA-L | |
10313134 | |
[Cl-].[Cl-].[Mn++] |
Cl2Mn | |
MFCD00011114 | |
manganese chloride, manganese ii chloride, manganese 2+ ion dichloride, manganesechloride, acmc-20akkt, manganous chloride,anhydrous, ksc171o6h, manganous chloride, anhydr, manganese ii chloride anhydrous crystalline | |
CHEBI:63041 |
Safety and Handling
GHS H Statement
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure if inhaled.
Causes serious eye damage.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Do not eat,drink or smoke when using th
GHS Signal Word: Danger
EINECSNumber : 231-869-6
RUO – Research Use Only