Learn More
Manganese(II) chloride tetrahydrate, 99+%, for analysis, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 205890025
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Manganese(II) chloride tetrahydrate | |
2.5kg | |
Cl2Mn·4H2O | |
07,222; 09,288 | |
Solubility in water: 1980g/L (20°C). Other solubilities: soluble in alcohol,insoluble in ether | |
197.91 | |
Analysis |
13446-34-9 | |
99% min. (Complexometry) | |
MnCl2·4H2O | |
15,5793 | |
197.91 | |
99+% | |
Plastic bottle |
Safety and Handling
GHS H Statement
May cause damage to organs through prolonged or repeated exposure if inhaled.
Harmful if swallowed.
Toxic to aquatic life with long lasting effects.
Causes serious eye damage.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
RUO â Research Use Only