missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Manganese(II) nitrate tetrahydrate, for analysis, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 193460010

 View more versions of this product

Catalog No. AC193460010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Manganese(II) nitrate tetrahydrate
20694-39-7
Crystals
MnN2O6·4H2O
MFCD00149789
10, 5555
Solubility in water: 3800g/L (20°C)
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.[Mn+2]
251.01
251.01
Analytical
37.0°C
Pink
1 kg
Mn(NO3)2·4H2O
02,257
manganese ii nitrate tetrahydrate, manganese nitrate tetrahydrate, unii-03m0g68r5f, manganese 2+ ion tetrahydrate dinitrate, manganese dinitrate tetrahydrate, acmc-20akkk, nitric acid, manganese 2+ salt, tetrahydrate, ksc206g2f, mn.2no3.4h2o, manganese 2+ dinitrate tetrahydrate
ALIMWUQMDCBYFM-UHFFFAOYSA-N
manganese(2+);dinitrate;tetrahydrate
182616
≥97.5%
Plastic bottle

Chemical Identifiers

20694-39-7
251.01
ALIMWUQMDCBYFM-UHFFFAOYSA-N
182616
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.[Mn+2]
MnN2O6·4H2O
MFCD00149789
manganese ii nitrate tetrahydrate, manganese nitrate tetrahydrate, unii-03m0g68r5f, manganese 2+ ion tetrahydrate dinitrate, manganese dinitrate tetrahydrate, acmc-20akkk, nitric acid, manganese 2+ salt, tetrahydrate, ksc206g2f, mn.2no3.4h2o, manganese 2+ dinitrate tetrahydrate
manganese(2+);dinitrate;tetrahydrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
May intensify fire; oxidizer.
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause damage to organs through prolonged or repeated exposure if inhaled.
Corrosive to the respiratory tract.<

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or ha

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only