Learn More
Manganese(II) perchlorate hexahydrate, Reagent Grade, Thermo Scientific Chemicals
$82.93 - $347.40
Chemical Identifiers
CAS | 15364-94-0 |
---|---|
Molecular Formula | Cl2H12MnO14 |
Molecular Weight (g/mol) | 361.92 |
MDL Number | MFCD00150255 |
InChI Key | AIFGVOMWPFMOCN-UHFFFAOYSA-L |
Synonym | manganese perchlorate hexahydrate, manganese ii perchlorate hexahydrate, perchloric acid, manganese 2+ salt, hexahydrate, 2clo4.mn.6h2o, manganese 2+ diperchlorate hexahydrate, manganese 2+ ion hexahydrate diperchlorate, manganese ii hexahydrate diperchlorate ion, manganese ii perchlorate hexahydrate, reagent grade |
PubChem CID | 203855 |
IUPAC Name | manganese(2+);diperchlorate;hexahydrate |
SMILES | O.O.O.O.O.O.[Mn++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O |
Description
It used in rocket fuel. This is one of the most proven materials for etching of liquid crystal displays and critical electronics applications as well as ore extraction. It is also used for electropolishing/etching of aluminum, molybdenum, and other metals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
15364-94-0 | |
361.92 | |
AIFGVOMWPFMOCN-UHFFFAOYSA-L | |
203855 | |
O.O.O.O.O.O.[Mn++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O |
Cl2H12MnO14 | |
MFCD00150255 | |
manganese perchlorate hexahydrate, manganese ii perchlorate hexahydrate, perchloric acid, manganese 2+ salt, hexahydrate, 2clo4.mn.6h2o, manganese 2+ diperchlorate hexahydrate, manganese 2+ ion hexahydrate diperchlorate, manganese ii hexahydrate diperchlorate ion, manganese ii perchlorate hexahydrate, reagent grade | |
manganese(2+);diperchlorate;hexahydrate |
Specifications
165°C (decomposition) | |
Crystalline | |
Cl2H12MnO14 | |
UN1481 | |
Soluble in benzene, ethyl acetate | |
O.O.O.O.O.O.[Mn++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O | |
361.92 | |
361.93 (253.84 Anhydrous) | |
Odorless | |
Manganese(II) perchlorate hexahydrate |
15364-94-0 | |
100 g | |
MFCD00150255 | |
manganese perchlorate hexahydrate, manganese ii perchlorate hexahydrate, perchloric acid, manganese 2+ salt, hexahydrate, 2clo4.mn.6h2o, manganese 2+ diperchlorate hexahydrate, manganese 2+ ion hexahydrate diperchlorate, manganese ii hexahydrate diperchlorate ion, manganese ii perchlorate hexahydrate, reagent grade | |
AIFGVOMWPFMOCN-UHFFFAOYSA-L | |
manganese(2+);diperchlorate;hexahydrate | |
203855 | |
Reagent | |
Hygroscopic |
Safety and Handling
GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P220-P221-P280-P370+P378q-P501c
H272
DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.
EINECSNumber : 237-390-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only