missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Manganese(II) sulfide, 99.9% (metals basis), Thermo Scientific Chemicals
$74.88 - $924.04
Chemical Identifiers
CAS | 18820-29-6 |
---|---|
Molecular Formula | MnS |
Molecular Weight (g/mol) | 87.00 |
MDL Number | MFCD00014210 |
InChI Key | VCTOKJRTAUILIH-UHFFFAOYSA-N |
Synonym | manganese sulfide, manganese ii sulfide, manganous sulfide, manganese sulphide, manganese 2+ sulfide, 12687-82-0 cpd with unspecified mf, manganese ii sulfide, ∼100 mesh particle size |
PubChem CID | 87809 |
SMILES | [S--].[Mn++] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA1283506
|
Thermo Scientific Chemicals
01283506 |
5 g |
Each for $74.88
|
|
|||||
AA1283514
|
Thermo Scientific Chemicals
01283514 |
25 g |
Each for $279.41
|
|
|||||
AA1283522
|
Thermo Scientific Chemicals
01283522 |
100 g |
Each for $924.04
|
|
|||||
Description
it is used as an intermediate in chemical research& pharmaceutical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
18820-29-6 | |
87.00 | |
VCTOKJRTAUILIH-UHFFFAOYSA-N | |
87809 |
MnS | |
MFCD00014210 | |
manganese sulfide, manganese ii sulfide, manganous sulfide, manganese sulphide, manganese 2+ sulfide, 12687-82-0 cpd with unspecified mf, manganese ii sulfide, ∼100 mesh particle size | |
[S--].[Mn++] |
Specifications
1610°C (decomposition) | |
18820-29-6 | |
5 g | |
MnS | |
manganese sulfide, manganese ii sulfide, manganous sulfide, manganese sulphide, manganese 2+ sulfide, 12687-82-0 cpd with unspecified mf, manganese ii sulfide, ∼100 mesh particle size | |
VCTOKJRTAUILIH-UHFFFAOYSA-N | |
sulfanylidenemanganese | |
87809 | |
Green powder | |
Manganese(II) sulfide |
Green | |
Powder | |
99.9% (Metal basis) | |
MFCD00014210 | |
It is soluble in water. 0.47mg/100 mL (18°C). | |
[S--].[Mn++] | |
87.00 | |
87 | |
3.99 g/cm3 at 20°C |
Safety and Handling
H301: Toxic if swallowed.
EINECSNumber : 242-599-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only