missing translation for 'onlineSavingsMsg'
Learn More

Manganese(III) oxide, 98%

CAS: 1317-34-6 | Mn2O3 | 157.873 g/mol

$330.11 - $1244.53

Chemical Identifiers

CAS 1317-34-6
Molecular Formula Mn2O3
Molecular Weight (g/mol) 157.873
MDL Number MFCD00016217
InChI Key GEYXPJBPASPPLI-UHFFFAOYSA-N
Synonym manganese iii oxide, manganic oxide, dimanganese trioxide, manganese sesquioxide, manganese trioxide, manganese manganate, manganese sisquioxide, manganese 3+ oxide, manganese oxide, oxo oxomanganiooxy manganese
PubChem CID 14824
IUPAC Name oxo(oxomanganiooxy)manganese
SMILES O=[Mn]O[Mn]=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA8779136
View Documents
Thermo Scientific Chemicals
08779136
500 g
Each for $330.11
Only null left
 
AA87791A3
View Documents
Thermo Scientific Chemicals
087791A3
2 kg
Each for $1,244.53
Only null left
 
Description

Description

Manganese(III) oxide is used to produce ceramic magnets and semiconductors. Further, it serves as a precursor to lithium manganese oxide. In addition to this, it acts as a useful battery cathode.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1317-34-6
157.873
GEYXPJBPASPPLI-UHFFFAOYSA-N
14824
O=[Mn]O[Mn]=O
Mn2O3
MFCD00016217
manganese iii oxide, manganic oxide, dimanganese trioxide, manganese sesquioxide, manganese trioxide, manganese manganate, manganese sisquioxide, manganese 3+ oxide, manganese oxide, oxo oxomanganiooxy manganese
oxo(oxomanganiooxy)manganese

Specifications

1080°C (decomposition)
Powder
98%
MFCD00016217
manganese iii oxide, manganic oxide, dimanganese trioxide, manganese sesquioxide, manganese trioxide, manganese manganate, manganese sisquioxide, manganese 3+ oxide, manganese oxide, oxo oxomanganiooxy manganese
GEYXPJBPASPPLI-UHFFFAOYSA-N
oxo(oxomanganiooxy)manganese
14824
98%
Manganese(III) oxide
1317-34-6
500 g
Mn2O3
14,5737
Soluble in acid and ammonium chloride. Insoluble in water,alcohol and acetone.
O=[Mn]O[Mn]=O
157.873
157.86
4.50 g/mL
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H332-H315-H319-H335

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P362-P301+P312-P312-P405-P403+P233-P501a

Warning

EINECSNumber : 215-264-4

RTECSNumber : OP0915000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only