Learn More
Manganese(IV) oxide, 80-85%, powder, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 213490010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentSpecifications
Manganese(IV) oxide | |
1313-13-9 | |
Powder | |
80-85% | |
MnO2 | |
01,637; 02,257; 03,191; 04,317; 05,422; 06,357; 08,312; 13,233; 14,200; 15,197; 16,202 | |
manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein | |
NUJOXMJBOLGQSY-UHFFFAOYSA-N | |
dioxomanganese | |
14801 | |
80 to 85% | |
Plastic bottle |
535.0°C | |
Gray | |
1 kg | |
MnO2 | |
MFCD00003463 | |
15, 5795 | |
Solubility in water: insoluble. Other solubilities: soluble in hydrochloric acid | |
O=[Mn]=O | |
86.94 | |
86.94 | |
0.5% max. (105°C) | |
5.2g/mL |
Chemical Identifiers
1313-13-9 | |
86.94 | |
NUJOXMJBOLGQSY-UHFFFAOYSA-N | |
14801 | |
O=[Mn]=O |
MnO2 | |
MFCD00003463 | |
manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein | |
dioxomanganese |
Safety and Handling
GHS H Statement
Harmful if inhaled.
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure if inhaled.
GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
D
GHS Signal Word: Warning
EINECSNumber : 215-202-6
RUO – Research Use Only