missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Manganese(IV) oxide, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A10765A9

 View more versions of this product

Catalog No. AAA10765A9


Explore available promotions

Description

Description

Manganese(IV) oxide is used as a pigment and as a reagent inorganic synthesis especially for the oxidation of allylic alcohols. It is also used for dry -cell batteries such as the alkaline battery and the zinc-carbon battery. It is involved in the preparation of other manganese compounds. It is a precursor to ferromanganese and related alloys, which is useful in the steel industry. It catalyses the decomposition of hydrogen peroxide into oxygen and water. Further, it is used to prepare manganese(II) sulfate, manganese(II) chloride and potassium permanganate. In addition to this, it is used in ceramics and glass manufacturing as an inorganic pigment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Manganese(IV) oxide is used as a pigment and as a reagent inorganic synthesis especially for the oxidation of allylic alcohols. It is also used for dry -cell batteries such as the alkaline battery and the zinc-carbon battery. It is involved in the preparation of other manganese compounds. It is a precursor to ferromanganese and related alloys, which is useful in the steel industry. It catalyses the decomposition of hydrogen peroxide into oxygen and water. Further, it is used to prepare manganese(II) sulfate, manganese(II) chloride and potassium permanganate. In addition to this, it is used in ceramics and glass manufacturing as an inorganic pigment.

Solubility
Insoluble in water.

Notes
Incompatible with strong acids, strong reducing agents and organic materials.
Specifications

Specifications

Manganese (IV) oxide
1313-13-9
MnO2
14,5730
Insoluble in water.
O=[Mn]=O
86.94
86.94
5.026
535°C (decomposition)
10 kg
MFCD00003463
manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein
NUJOXMJBOLGQSY-UHFFFAOYSA-N
dioxomanganese
14801
98%

Chemical Identifiers

1313-13-9
86.94
NUJOXMJBOLGQSY-UHFFFAOYSA-N
14801
O=[Mn]=O
MnO2
MFCD00003463
manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein
dioxomanganese
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.

P260-P264b-P270-P271-P301+P312-P304+P340-P312-P314-P330-P501c

H302+H332-H373

EINECSNumber : 215-202-6

RTECSNumber : OP0350000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only