missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Manganese pieces, irregular, 99.9% (metals basis), Thermo Scientific Chemicals

$103.55 - $529.59

Chemical Identifiers

CAS 7439-96-5
Molecular Formula Mn
Molecular Weight (g/mol) 54.94
MDL Number MFCD00011111
InChI Key PWHULOQIROXLJO-UHFFFAOYSA-N
Synonym colloidal, mangan, manganese, elemental, cutaval, magnacat, metal alloy, tronamang, fume, mangan polish, compounds
PubChem CID 23930
ChEBI CHEBI:35154
IUPAC Name manganese
SMILES [Mn]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1023630
View Documents
Thermo Scientific Chemicals
01023630
250 g
Each for $103.55
 
AA10236A1
View Documents
Thermo Scientific Chemicals
010236A1
1 kg
Each for $230.10
 
AA10236A7
View Documents
Thermo Scientific Chemicals
10236A7
5 kg
Each for $529.59
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7439-96-5
54.94
PWHULOQIROXLJO-UHFFFAOYSA-N
23930
manganese
Mn
MFCD00011111
colloidal, mangan, manganese, elemental, cutaval, magnacat, metal alloy, tronamang, fume, mangan polish, compounds
CHEBI:35154
[Mn]

Specifications

Gray
Gray irregularly shaped pieces
99.9% (Metals basis)
MFCD00011111
colloidal, mangan, manganese, elemental, cutaval, magnacat, metal alloy, tronamang, fume, mangan polish, compounds
[Mn]
54.94
CHEBI:35154
Broken Plate
7439-96-5
7.3g/cm3 at 20°C
Mn
250 g
PWHULOQIROXLJO-UHFFFAOYSA-N
manganese
23930
54.94
Manganese
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-105-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only