missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Manganese powder, -325 mesh, 99.95% (metals basis), Thermo Scientific Chemicals

$489.41 - $489.41

Chemical Identifiers

CAS 7439-96-5
Molecular Formula Mn
Molecular Weight (g/mol) 54.94
MDL Number MFCD00011111
InChI Key PWHULOQIROXLJO-UHFFFAOYSA-N
Synonym colloidal, mangan, manganese, elemental, cutaval, magnacat, metal alloy, tronamang, fume, mangan polish, compounds
PubChem CID 23930
ChEBI CHEBI:35154
IUPAC Name manganese
SMILES [Mn]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3631522
View Documents
Thermo Scientific Chemicals
03631522
100 g
Each for $489.41
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7439-96-5
54.94
PWHULOQIROXLJO-UHFFFAOYSA-N
23930
manganese
Mn
MFCD00011111
colloidal, mangan, manganese, elemental, cutaval, magnacat, metal alloy, tronamang, fume, mangan polish, compounds
CHEBI:35154
[Mn]

Specifications

7439-96-5
99.95%, metal basis
MFCD00011111
colloidal, mangan, manganese, elemental, cutaval, magnacat, metal alloy, tronamang, fume, mangan polish, compounds
[Mn]
54.94
CHEBI:35154
Powder
7.3g/cm3 at 20°C
Mn
100 g
PWHULOQIROXLJO-UHFFFAOYSA-N
manganese
23930
-325 Mesh
Manganese
Videos
Safety and Handling

Safety and Handling

Take precautionary measures against static discharges.

P210-P240-P241-P264b-P280-P305+P351+P338-P370+P378q

H228-H319

DOTInformation : DOT Class: 4.1; Packaging Group: III

EINECSNumber : 231-105-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only