missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Manganese Sulfate Monohydrate (Crystalline/Certified ACS), Fisher Chemical™

Supplier:  Thermo Fisher Scientific M1133

 View more versions of this product

Catalog No. M113-3



Explore available promotions

Description

Description

Specifications

Specifications

850°C
700°C
Pale Pink
3 kg
H2MnO5S
manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate
O.[Mn++].[O-]S([O-])(=O)=O
169.01
CHEBI:86364
98.0 to 101.0%
Pass Test
Poly Bottle
3.25g/cm³
Manganese Sulfate Monohydrate
10034-96-5
Solid
98.0 to 101.0 %
MFCD00149159
ISPYRSDWRDQNSW-UHFFFAOYSA-L
manganese(2+) hydrate sulfate
177577
169.02
Certified ACS
10 to 12%
850°C

Chemical Identifiers

10034-96-5
169.01
ISPYRSDWRDQNSW-UHFFFAOYSA-L
177577
manganese(2+) hydrate sulfate
H2MnO5S
MFCD00149159
manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate
CHEBI:86364
O.[Mn++].[O-]S([O-])(=O)=O
Videos
Safety and Handling

Safety and Handling

SDS Documents

Documents

Promotions

Promotions