missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Manganese Sulfate Monohydrate (Crystalline/Certified ACS), Fisher Chemical™
Supplier: Thermo Fisher Scientific M113500
Specifications
850°C | |
700°C | |
Pale Pink | |
500 g | |
H2MnO5S | |
manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate | |
O.[Mn++].[O-]S([O-])(=O)=O | |
169.01 | |
CHEBI:86364 | |
98.0 to 101.0% | |
Pass Test | |
Poly Bottle | |
3.25g/cm³ |
Manganese Sulfate Monohydrate | |
10034-96-5 | |
Solid | |
98.0 to 101.0 % | |
MFCD00149159 | |
ISPYRSDWRDQNSW-UHFFFAOYSA-L | |
manganese(2+) hydrate sulfate | |
177577 | |
169.02 | |
Certified ACS | |
10 to 12% | |
850°C |
Chemical Identifiers
10034-96-5 | |
169.01 | |
ISPYRSDWRDQNSW-UHFFFAOYSA-L | |
177577 | |
manganese(2+) hydrate sulfate |
H2MnO5S | |
MFCD00149159 | |
manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate | |
CHEBI:86364 | |
O.[Mn++].[O-]S([O-])(=O)=O |