missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Manganese Sulfate Monohydrate, (Crystalline),MP Biomedicals

Supplier:  MP Biomedicals Inc 0219470280

 View more versions of this product

Catalog No. ICN19470280

Explore available promotions

Description

Description

Specifications

Specifications

Manganese Sulfate Monohydrate
Crystalline
H2MnO5S
manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate
O.[Mn++].[O-]S([O-])(=O)=O
169.01
CHEBI:86364
10034-96-5
100 g
MFCD00149159
ISPYRSDWRDQNSW-UHFFFAOYSA-L
manganese(2+) hydrate sulfate
177577
169

Chemical Identifiers

10034-96-5
169.01
ISPYRSDWRDQNSW-UHFFFAOYSA-L
177577
manganese(2+) hydrate sulfate
H2MnO5S
MFCD00149159
manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate
CHEBI:86364
O.[Mn++].[O-]S([O-])(=O)=O
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Room Temperature

SDS Documents

Documents

Promotions

Promotions