missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Mannose triflate, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 461341000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Mannose triflate | |
Authentic | |
C15H19F3O12S | |
tatm, mannose triflate, 1,3,4,6-tetra-o-acetyl-2-o-trifluoromethanesulfonyl-beta-d-mannopyranose, 1,3,4,6-tetra-o-acetyl-2-o-trifluoromethanesulfonyl-b-d-mannopyranose, unii-0ki8ffq6tt, 0ki8ffq6tt, beta-d-mannopyranose 1,3,4,6-tetra-o-acetate 2-o-trifluoromethanesulfonate, 2r,3r,4s,5s,6s-3,4,6-tris acetyloxy-5-trifluoromethanesulfonyloxy oxan-2-yl methyl acetate, mannose trilfate | |
OIBDVHSTOUGZTJ-PEBLQZBPSA-N | |
[(2R,3R,4S,5S,6S)-3,4,6-triacetyloxy-5-(trifluoromethylsulfonyloxy)oxan-2-yl]methyl acetate | |
11113597 | |
98% |
92051-23-5 | |
97.5% min. (ELSD) (HPLC) | |
100mg | |
Clear colorless to slightly yellow | |
CC(=O)OCC1C(C(C(C(O1)OC(=O)C)OS(=O)(=O)C(F)(F)F)OC(=O)C)OC(=O)C | |
480.36 | |
480.36 |
RUO â Research Use Only