missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Manoalide, 98%, Thermo Scientific Chemicals

 

Supplier:  Thermo Scientific Chemicals J67341MCR

Catalog No. AAJ67341MCR


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Manoalide
Colorless to White
MFCD00153826
manoalide, unii-e1dk0157k9, 2 5h-furanone, 4-2r,6r-3,6-dihydro-6-hydroxy-5-3e-4-methyl-6-2,6,6-trimethyl-1-cyclohexen-1-yl-3-hexenyl-2h-pyran-2-yl-5-hydroxy-, 5r, 2 5h-furanone, 4-3,6-dihydro-6-hydroxy-5-4-methyl-6-2,6,6-trimethyl-1-cyclohexen-1-yl-3-hexenyl-2h-pyran-2-yl-5-hydroxy, 5-hydroxy-4-6-hydroxy-5-e-4-methyl-6-2,6,6-trimethyl-cyclohex-1-enyl-hex-3-enyl-3,6-dihydro-2h-pyran-2-yl-5h-furan-2-one, 2r-2-hydroxy-3-2r,6r-6-hydroxy-5-e-4-methyl-6-2,6,6-trimethyl-1-cyclohexenyl hex-3-enyl-3,6-dihydro-2h-pyran-2-yl-2h-furan-5-one, 2r-2-hydroxy-3-2r,6r-6-hydroxy-5-e-4-methyl-6-2,6,6-trimethylcyclohexen-1-yl hex-3-enyl-3,6-dihydro-2h-pyran-2-yl-2h-furan-5-one, 2r-3-2r,6r-5-e-4-methyl-6-2,6,6-trimethylcyclohexen-1-yl hex-3-enyl-6-oxidanyl-3,6-dihydro-2h-pyran-2-yl-2-oxidanyl-2h-furan-5-one, 5r-5-hydroxy-4-2r,6r-6-hydroxy-5-3e-4-methyl-6-2,6,6-trimethylcyclohex-1-en-1-yl hex-3-en-1-yl-3,6-dihydro-2h-pyran-2-yl furan-2 5h-one, 2 5h-furanone,4-2r,6r-3,6-dihydro-6-hydroxy-5-3e-4-methyl-6-2,6,6-trimethyl-1-cyclohexen-1-yl-3-hexen-1-yl-2h-pyran-2-yl-5-hydroxy-, 5r
CC1=C(C(CCC1)(C)C)CCC(=CCCC2=CCC(OC2O)C3=CC(=O)OC3O)C
416.558
CHEBI:66666
98%
75088-80-1
C25H36O5
1 mg
FGJIDQWRRLDGDB-CPIXEKRISA-N
(2R)-2-hydroxy-3-[(2R,6R)-6-hydroxy-5-[(E)-4-methyl-6-(2,6,6-trimethylcyclohexen-1-yl)hex-3-enyl]-3,6-dihydro-2H-pyran-2-yl]-2H-furan-5-one
6437368
416.56
Solid

Chemical Identifiers

75088-80-1
416.558
FGJIDQWRRLDGDB-CPIXEKRISA-N
6437368
(2R)-2-hydroxy-3-[(2R,6R)-6-hydroxy-5-[(E)-4-methyl-6-(2,6,6-trimethylcyclohexen-1-yl)hex-3-enyl]-3,6-dihydro-2H-pyran-2-yl]-2H-furan-5-one
C25H36O5
MFCD00153826
manoalide, unii-e1dk0157k9, 2 5h-furanone, 4-2r,6r-3,6-dihydro-6-hydroxy-5-3e-4-methyl-6-2,6,6-trimethyl-1-cyclohexen-1-yl-3-hexenyl-2h-pyran-2-yl-5-hydroxy-, 5r, 2 5h-furanone, 4-3,6-dihydro-6-hydroxy-5-4-methyl-6-2,6,6-trimethyl-1-cyclohexen-1-yl-3-hexenyl-2h-pyran-2-yl-5-hydroxy, 5-hydroxy-4-6-hydroxy-5-e-4-methyl-6-2,6,6-trimethyl-cyclohex-1-enyl-hex-3-enyl-3,6-dihydro-2h-pyran-2-yl-5h-furan-2-one, 2r-2-hydroxy-3-2r,6r-6-hydroxy-5-e-4-methyl-6-2,6,6-trimethyl-1-cyclohexenyl hex-3-enyl-3,6-dihydro-2h-pyran-2-yl-2h-furan-5-one, 2r-2-hydroxy-3-2r,6r-6-hydroxy-5-e-4-methyl-6-2,6,6-trimethylcyclohexen-1-yl hex-3-enyl-3,6-dihydro-2h-pyran-2-yl-2h-furan-5-one, 2r-3-2r,6r-5-e-4-methyl-6-2,6,6-trimethylcyclohexen-1-yl hex-3-enyl-6-oxidanyl-3,6-dihydro-2h-pyran-2-yl-2-oxidanyl-2h-furan-5-one, 5r-5-hydroxy-4-2r,6r-6-hydroxy-5-3e-4-methyl-6-2,6,6-trimethylcyclohex-1-en-1-yl hex-3-en-1-yl-3,6-dihydro-2h-pyran-2-yl furan-2 5h-one, 2 5h-furanone,4-2r,6r-3,6-dihydro-6-hydroxy-5-3e-4-methyl-6-2,6,6-trimethyl-1-cyclohexen-1-yl-3-hexen-1-yl-2h-pyran-2-yl-5-hydroxy-, 5r
CHEBI:66666
CC1=C(C(CCC1)(C)C)CCC(=CCCC2=CCC(OC2O)C3=CC(=O)OC3O)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only