Stoelting™ Manual Stereotaxic Injector
Stoelting Manual Stereotaxic Injector provides an economical means of injecting/withdrawing a calibrated volume with simple, precise manual control.
Supplier:  Stoelting™ 51639
Catalog No. 10000164
Explore available promotions
Description
Description
  • Mounted directly on a stereotaxic frame
  • Allows precise injection directly through the needle into the tissue.
  • Avoids compliance issues that arise when injections are forced through plastic tubing.
  • Volumes as small as 200 picoliters may be infused or withdrawn.
Specifications
Specifications
Neuroscience