MAPA™ Kroflex 840 Cut-Resistant Gloves, Latex Coated
Enhanced non-slip feature offers ultra-protection when handling heavy/sharp objects in wet environments.
Supplier:  MAPA™ 34840029
 View more versions of this product
Catalog No. 19-249-757
Explore available promotions
Alert:
This product contains natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Description
Description
Ideal for use when handling glass sheets, hot plastic parts, composite materials
  • Natural rubber crinkle palm coating
  • Mechanical durability
  • Anatomical shape and flexibility
  • Comfortable for extended wear, with a secure grip
Specifications
Specifications
ANSI 2
Latex
ANSI A4
HDPE
9
Handling Glass, hot Plastic parts and waste treatment applications
840419
10.5 in.
EN 421 , EN 407 , EN 511 , CE 3543
Blue, Gray
No
Cut-resistant Gloves
EN 388
Knit
Kroflex 840
26.5 cm