missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MAPA™ ChemZoil™ NL339 Chemical Resistant Neoprene Gloves
Neoprene multi-layer gloves provide chemical resistance against acids and alkalis, such as concentrated hydrofluoric acid, oils, and greases.
Supplier: MAPA™ 339429
Description
- Protects against acids
- For use with chemical protective suits
- Black, heavyweight, neoprene
- Articulated finger design for added comfort, flexibility and reduced hand fatigue
- Soft, interlocked knit cotton liner absorbs perspiration and adds comfort
- Multilayer construction assures a leakproof glove
- Roughened grip with pinked cuff
- Length: 14 in. (35.6 cm)
Specifications
21 CFR 170-199 | |
Black | |
Neoprene | |
50 mil | |
EN 388:3121 | |
ABCJKL | |
Chemical processing, alkaline units at petroleum refineries, metal plating operations, hazardous material handling, hazmat and chemical spills | |
Pebbled | |
Lined | |
ChemZoil™ NL-339 | |
35.5 cm |
Cat III CE 0334, EN 407:X1XXXX, EN 511 | |
Yes | |
9 | |
1.20 mm | |
Yes | |
Bacteria, Fungi only | |
Pinked | |
Cotton Knit | |
339429 | |
14 in. |